Order Entry
Canada
ContactUsLinkComponent
Anti-TNFSF13B Rabbit Recombinant Polyclonal Antibody (HRP)
Anti-TNFSF13B Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-TNFSF13B Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier:  AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 77808-910
  • 77808-911
  • Antigen symbol:
  • TNFSF13B
  • TNFSF13B
  • Antigen name:
  • Tumor necrosis factor ligand superfamily member 13B protein (68-285AA)
  • Tumor necrosis factor ligand superfamily member 13B protein (68-285AA)
  • Antigen synonyms:
  • ApoL related ligand TALL 1|B cell Activating Factor|B lymphocyte stimulator|B-cell-activating factor|BAFF|BLyS|CD 257|CD257|CD257 antigen|Delta BAFF|Dendritic cell derived TNF like molecule|Dendritic cell-derived TNF-like molecule|DTL|DTL precursor|PRO738|soluble form|TALL 1|TALL-1|TALL1|THANK|TN13B_HUMAN|TNF and APOL related leukocyte expressed ligand 1|TNF homolog that activates apoptosis|TNF homolog that activates apoptosis NKFB and JNK.|TNF- and APOL-related leukocyte expressed ligand 1|TNFSF13B|TNFSF20|TNLG7A|Tumor necrosis factor (ligand) superfamily member 13b|Tumor necrosis factor ligand 7A|Tumor necrosis factor ligand superfamily member 13b|Tumor necrosis factor ligand superfamily member 20|Tumor necrosis factor like protein ZTNF4|Tumor necrosis factor superfamily member 13B|UNQ401|ZTNF4
  • ApoL related ligand TALL 1|B cell Activating Factor|B lymphocyte stimulator|B-cell-activating factor|BAFF|BLyS|CD 257|CD257|CD257 antigen|Delta BAFF|Dendritic cell derived TNF like molecule|Dendritic cell-derived TNF-like molecule|DTL|DTL precursor|PRO738|soluble form|TALL 1|TALL-1|TALL1|THANK|TN13B_HUMAN|TNF and APOL related leukocyte expressed ligand 1|TNF homolog that activates apoptosis|TNF homolog that activates apoptosis NKFB and JNK.|TNF- and APOL-related leukocyte expressed ligand 1|TNFSF13B|TNFSF20|TNLG7A|Tumor necrosis factor (ligand) superfamily member 13b|Tumor necrosis factor ligand 7A|Tumor necrosis factor ligand superfamily member 13b|Tumor necrosis factor ligand superfamily member 20|Tumor necrosis factor like protein ZTNF4|Tumor necrosis factor superfamily member 13B|UNQ401|ZTNF4
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • HRP (Horseradish Peroxidase)
  • HRP (Horseradish Peroxidase)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9Y275
  • Q9Y275
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, protein G purified
  • >95%, protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together