Anti-PRDX2 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PRDX2
Clonality: Recombinant
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77816-242
- 77816-243
- Antigen symbol:
- PRDX2
- PRDX2
- Antigen name:
- Peroxiredoxin-2 protein (2-65AA)
- Peroxiredoxin-2 protein (2-65AA)
- Antigen synonyms:
- Epididymis secretory sperm binding protein Li 2a|HEL S 2a|MGC4104|Natural killer cell enhancing factor B|Natural killer cell-enhancing factor B|Natural Killer Enhancing Factor B|NKEF B|NKEF-B|NKEFB|Peroxiredoxin 2|Peroxiredoxin-2|PRDX 2|PRDX2|PRDX2_HUMAN|PrP|PRX2|PRXII|PTX1|TDPX1|Thiol Specific Antioxidant 1|Thiol specific antioxidant protein|Thiol-specific antioxidant protein|Thioredoxin Dependent Peroxide Reductase 1|Thioredoxin peroxidase 1|Thioredoxin-dependent peroxide reductase 1|Torin|TPX1|TSA
- Epididymis secretory sperm binding protein Li 2a|HEL S 2a|MGC4104|Natural killer cell enhancing factor B|Natural killer cell-enhancing factor B|Natural Killer Enhancing Factor B|NKEF B|NKEF-B|NKEFB|Peroxiredoxin 2|Peroxiredoxin-2|PRDX 2|PRDX2|PRDX2_HUMAN|PrP|PRX2|PRXII|PTX1|TDPX1|Thiol Specific Antioxidant 1|Thiol specific antioxidant protein|Thiol-specific antioxidant protein|Thioredoxin Dependent Peroxide Reductase 1|Thioredoxin peroxidase 1|Thioredoxin-dependent peroxide reductase 1|Torin|TPX1|TSA
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P32119
- P32119
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together