Anti-SPTLC1 Rabbit Polyclonal Antibody
77800-944
New Product
- Antigen name:HSAN
- Antigen symbol:SPTLC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O15269
- Antigen synonyms:HSAN|HSAN1|HSN1|LBC1|LCB 1|LCB1|Long chain base biosynthesis protein 1|MGC14645|Serine C palmitoyltransferase|Serine palmitoyl CoA transferase 1|Serine palmitoyltransferase 1|Serine palmitoyltransferase long chain base subunit 1|Serine palmitoyltransferase subunit 1|Serine-palmitoyl-CoA transferase 1|SPT 1|SPT1|SPTC1_HUMAN|SPTI|SPTLC 1|SPTLC1
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: SPTLC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat