Anti-DLG2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: DLG2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77824-538
- 77824-537
- Antigen symbol:
- DLG2
- DLG2
- Antigen name:
- Disks large homolog 2 protein (310-420AA)
- Disks large homolog 2 protein (310-420AA)
- Antigen synonyms:
- Channel associated protein of synapse 110|Channel associated protein of synapses 110kD|Channel-associated protein of synapse-110|Chapsyn 110|Chapsyn-110|Chapsyn110|Discs|large homolog 2 (Drosophila)|Disks large homolog 2|DKFZp781D1854|DKFZp781E0954|Dlg 2|dlg2|DLG2_HUMAN|Dlgh 2|Dlgh2|FLJ37266|Gm1197|MGC131811|Postsynaptic density protein PSD 93|Postsynaptic density protein PSD-93|Postsynaptic density protein PSD93|PSD 93|PSD93
- Channel associated protein of synapse 110|Channel associated protein of synapses 110kD|Channel-associated protein of synapse-110|Chapsyn 110|Chapsyn-110|Chapsyn110|Discs|large homolog 2 (Drosophila)|Disks large homolog 2|DKFZp781D1854|DKFZp781E0954|Dlg 2|dlg2|DLG2_HUMAN|Dlgh 2|Dlgh2|FLJ37266|Gm1197|MGC131811|Postsynaptic density protein PSD 93|Postsynaptic density protein PSD-93|Postsynaptic density protein PSD93|PSD 93|PSD93
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15700
- Q15700
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together