Anti-PIK3R6 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PIK3R6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77828-248
- 77828-249
- Antigen symbol:
- C17orf38;Chromosome 17 open reading frame 38;HsT41028;p84 PI3K adapter protein;p84 PIKAP;p87 phosphoinositide 3-kinase gamma (PI3Kg) adapter protein;p87 PI3K adapter protein;p87PIKAP;Phosphatidylinositol 3-kinase;regulatory subunit 6;Phosphoinositide 3-kinase gamma adapter protein of 87 kDa;Phosphoinositide 3-kinase regulatory subunit 6;PI3Kgamma adapter protein of 87 kDa;PI3R6_HUMAN;PIK3R6
- C17orf38;Chromosome 17 open reading frame 38;HsT41028;p84 PI3K adapter protein;p84 PIKAP;p87 phosphoinositide 3-kinase gamma (PI3Kg) adapter protein;p87 PI3K adapter protein;p87PIKAP;Phosphatidylinositol 3-kinase;regulatory subunit 6;Phosphoinositide 3-kinase gamma adapter protein of 87 kDa;Phosphoinositide 3-kinase regulatory subunit 6;PI3Kgamma adapter protein of 87 kDa;PI3R6_HUMAN;PIK3R6
- Antigen name:
- Phosphoinositide 3-kinase regulatory subunit 6 protein (566-667AA)
- Phosphoinositide 3-kinase regulatory subunit 6 protein (566-667AA)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5UE93
- Q5UE93
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together