Anti-RPLP1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: RPLP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77802-793
- 77802-794
- Antigen symbol:
- RPLP1
- RPLP1
- Antigen name:
- 60S acidic ribosomal protein P1 protein (2-114AA)
- 60S acidic ribosomal protein P1 protein (2-114AA)
- Antigen synonyms:
- 60S acidic ribosomal protein P1|RLA1_HUMAN|Rplp1
- 60S acidic ribosomal protein P1|RLA1_HUMAN|Rplp1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P05386
- P05386
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together