Anti-CXXC5 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: CXXC5
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77829-614
- 77829-615
- Antigen symbol:
- CXXC5
- CXXC5
- Antigen name:
- CXXC-type zinc finger protein 5 protein (1-227AA)
- CXXC-type zinc finger protein 5 protein (1-227AA)
- Antigen synonyms:
- CF5| CXXC finger 5 protein| CXXC finger protein 5| CXXC-type zinc finger protein 5| CXXC5| CXXC5_HUMAN| HSPC195| Putative MAPK-activating protein PM08| Putative NF-kappa-B-activating protein 102| retinoid-inducible nuclear factor| RINF| TCCCIA00297| WID| WT1-induced Inhibitor of Dishevelled
- CF5| CXXC finger 5 protein| CXXC finger protein 5| CXXC-type zinc finger protein 5| CXXC5| CXXC5_HUMAN| HSPC195| Putative MAPK-activating protein PM08| Putative NF-kappa-B-activating protein 102| retinoid-inducible nuclear factor| RINF| TCCCIA00297| WID| WT1-induced Inhibitor of Dishevelled
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7LFL8
- Q7LFL8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together