Anti-ITIH5 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ITIH5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77831-739
- 77831-738
- Antigen symbol:
- ITIH5
- ITIH5
- Antigen name:
- Inter-alpha-trypsin inhibitor heavy chain H5 protein (35-161AA)
- Inter-alpha-trypsin inhibitor heavy chain H5 protein (35-161AA)
- Antigen synonyms:
- ITIH5|KIAA1953|PP14776|UNQ311/PRO354|Inter-alpha-trypsin inhibitor heavy chain H5|ITI heavy chain H5|ITI-HC5|Inter-alpha-inhibitor heavy chain 5
- ITIH5|KIAA1953|PP14776|UNQ311/PRO354|Inter-alpha-trypsin inhibitor heavy chain H5|ITI heavy chain H5|ITI-HC5|Inter-alpha-inhibitor heavy chain 5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86UX2
- Q86UX2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together