Anti-TRIM9 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TRIM9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-438
- 77840-439
- Antigen symbol:
- TRIM9
- TRIM9
- Antigen name:
- E3 ubiquitin-protein ligase TRIM9 protein (1-550AA)
- E3 ubiquitin-protein ligase TRIM9 protein (1-550AA)
- Antigen synonyms:
- TRIM9|KIAA0282|RNF91|E3 ubiquitin-protein ligase TRIM9|EC 2.3.2.27|RING finger protein 91|RING-type E3 ubiquitin transferase TRIM9|Tripartite motif-containing protein 9
- TRIM9|KIAA0282|RNF91|E3 ubiquitin-protein ligase TRIM9|EC 2.3.2.27|RING finger protein 91|RING-type E3 ubiquitin transferase TRIM9|Tripartite motif-containing protein 9
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9C026
- Q9C026
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together