Anti-INCA1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: INCA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77821-631
- 77821-630
- Antigen symbol:
- INCA1
- INCA1
- Antigen name:
- Protein INCA1 protein (1-236AA)
- Protein INCA1 protein (1-236AA)
- Antigen synonyms:
- HSD45|Inca1|INCA1_HUMAN|Inhibitor of CDK cyclin A1 interacting protein 1|Inhibitor of CDK interacting with cyclin A1|MGC148150|MGC148151|Protein INCA1
- HSD45|Inca1|INCA1_HUMAN|Inhibitor of CDK cyclin A1 interacting protein 1|Inhibitor of CDK interacting with cyclin A1|MGC148150|MGC148151|Protein INCA1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q0VD86
- Q0VD86
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together