Order Entry
Canada
ContactUsLinkComponent
Anti-AMIGO2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Anti-AMIGO2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-AMIGO2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77831-883
  • 77831-884
  • Antigen symbol:
  • AMIGO2
  • AMIGO2
  • Antigen name:
  • Amphoterin-induced protein 2 protein (420-522AA)
  • Amphoterin-induced protein 2 protein (420-522AA)
  • Antigen synonyms:
  • Adhesion molecule with Ig like domain 2|ALI 1|ALI1|Alivin 1|Alivin-1|Alivin1|AMGO2_HUMAN|AMIGO 2|AMIGO-2|Amigo2|Amphoterin induced gene 2|Amphoterin induced protein 2|Amphoterin-induced gene and open reading frame 2|Amphoterin-induced protein 2|DEGA|Differentially expressed in gastric adenocarcinoma|Differentially expressed in gastric adenocarcinomas|Transmembrane protein AMIGO 2|Transmembrane protein AMIGO2
  • Adhesion molecule with Ig like domain 2|ALI 1|ALI1|Alivin 1|Alivin-1|Alivin1|AMGO2_HUMAN|AMIGO 2|AMIGO-2|Amigo2|Amphoterin induced gene 2|Amphoterin induced protein 2|Amphoterin-induced gene and open reading frame 2|Amphoterin-induced protein 2|DEGA|Differentially expressed in gastric adenocarcinoma|Differentially expressed in gastric adenocarcinomas|Transmembrane protein AMIGO 2|Transmembrane protein AMIGO2
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q86SJ2
  • Q86SJ2
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together