Anti-F11R Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:50 to 1:200
Type:
Antigen: F11R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77837-760
- 77837-759
- Antigen symbol:
- F11R
- F11R
- Antigen name:
- Fusion protein of F11R
- Fusion protein of F11R
- Antigen synonyms:
- CD 321|CD321|CD321 antigen|ESTM33|F11 receptor|F11R|JAM 1|JAM A|JAM|JAM-1|JAM-A|JAM1|JAM1_HUMAN|JAMA|JCAM|Jcam1|Junction adhesion molecule 1|Junction adhesion molecule|mouse|homolog of|Junctional adhesion molecule 1|Junctional adhesion molecule A|KAT|Ly106|PAM 1|PAM-1|PAM1|Platelet adhesion molecule 1|Platelet adhesion molecule|Platelet F11 receptor|PRO301|UNQ264
- CD 321|CD321|CD321 antigen|ESTM33|F11 receptor|F11R|JAM 1|JAM A|JAM|JAM-1|JAM-A|JAM1|JAM1_HUMAN|JAMA|JCAM|Jcam1|Junction adhesion molecule 1|Junction adhesion molecule|mouse|homolog of|Junctional adhesion molecule 1|Junctional adhesion molecule A|KAT|Ly106|PAM 1|PAM-1|PAM1|Platelet adhesion molecule 1|Platelet adhesion molecule|Platelet F11 receptor|PRO301|UNQ264
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y624
- Q9Y624
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together