Anti-PRDM2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PRDM2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77821-817
- 77821-816
- Antigen symbol:
- PRDM2
- PRDM2
- Antigen name:
- PR domain zinc finger protein 2 protein (733-961AA)
- PR domain zinc finger protein 2 protein (733-961AA)
- Antigen synonyms:
- GATA-3-binding protein G3B|HUMHOXY1|KMT8|Lysine N-methyltransferase 8|MTB-ZF|MTE-binding protein|PR domain containing 2|with ZNF domain|PR domain zinc finger protein 2|PR domain-containing protein 2|PRDM2|PRDM2_HUMAN|Retinoblastoma protein binding zinc finger protein|Retinoblastoma protein-interacting zinc finger protein|Retinoblastoma protein-interacting zinc-finger protein|RIZ|RIZ1|RIZ2|Zinc finger DNA binding protein|Zinc finger protein RIZ
- GATA-3-binding protein G3B|HUMHOXY1|KMT8|Lysine N-methyltransferase 8|MTB-ZF|MTE-binding protein|PR domain containing 2|with ZNF domain|PR domain zinc finger protein 2|PR domain-containing protein 2|PRDM2|PRDM2_HUMAN|Retinoblastoma protein binding zinc finger protein|Retinoblastoma protein-interacting zinc finger protein|Retinoblastoma protein-interacting zinc-finger protein|RIZ|RIZ1|RIZ2|Zinc finger DNA binding protein|Zinc finger protein RIZ
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13029
- Q13029
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together