Anti-PSORS1C1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: PSORS1C1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77841-515
- 77841-516
- Antigen symbol:
- PSORS1C1
- PSORS1C1
- Antigen name:
- Psoriasis susceptibility 1 candidate gene 1 protein (1-152AA)
- Psoriasis susceptibility 1 candidate gene 1 protein (1-152AA)
- Antigen synonyms:
- PSORS1C1|C6orf16|SEEK1|Psoriasis susceptibility 1 candidate gene 1 protein|Protein SEEK1
- PSORS1C1|C6orf16|SEEK1|Psoriasis susceptibility 1 candidate gene 1 protein|Protein SEEK1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UIG5
- Q9UIG5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together