Anti-SEMA3F Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SEMA3F
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-735
- 77824-734
- Antigen symbol:
- SEMA3F
- SEMA3F
- Antigen name:
- Semaphorin-3F protein (565-651AA)
- Semaphorin-3F protein (565-651AA)
- Antigen synonyms:
- SEM3F_HUMAN| sema domain immunoglobulin domain Ig short basic domain secreted 3F| sema domain immunoglobulin domain Ig short basic domain secreted semaphorin 3F| Sema III/F| Sema IV| SEMA-IV| SEMA3F| SEMA4| SemaIII/F| SemaIV| SEMAK| Semaphorin IV| Semaphorin-3F| Semaphorin3F| SemaphorinIV
- SEM3F_HUMAN| sema domain immunoglobulin domain Ig short basic domain secreted 3F| sema domain immunoglobulin domain Ig short basic domain secreted semaphorin 3F| Sema III/F| Sema IV| SEMA-IV| SEMA3F| SEMA4| SemaIII/F| SemaIV| SEMAK| Semaphorin IV| Semaphorin-3F| Semaphorin3F| SemaphorinIV
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13275
- Q13275
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together