Anti-ARPIN Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ARPIN
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-609
- 77832-608
- Antigen symbol:
- ARPIN
- ARPIN
- Antigen name:
- Arpin protein (1-226AA)
- Arpin protein (1-226AA)
- Antigen synonyms:
- 2610034B18Rik|Actin related protein 2/3 complex inhibitor|Arp2/3 inhibition protein|ARPIN|C15orf38|Chromosome 15 open reading frame 38|CO038_HUMAN|Hypothetical protein LOC348110|MGC61550|RIKEN cDNA 2610034B18|UPF0552 protein C15orf38
- 2610034B18Rik|Actin related protein 2/3 complex inhibitor|Arp2/3 inhibition protein|ARPIN|C15orf38|Chromosome 15 open reading frame 38|CO038_HUMAN|Hypothetical protein LOC348110|MGC61550|RIKEN cDNA 2610034B18|UPF0552 protein C15orf38
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7Z6K5
- Q7Z6K5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together