Anti-RND3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: RND3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77797-431
- 77797-430
- Antigen symbol:
- RND3
- RND3
- Antigen name:
- Rho-related GTP-binding protein RhoE protein (152-239AA)
- Rho-related GTP-binding protein RhoE protein (152-239AA)
- Antigen synonyms:
- ARHE|memB|Protein MemB|Ras homolog gene family member E|Rho family GTPase 3|Rho-related GTP-binding protein Rho8|Rho-related GTP-binding protein RhoE|Rho8|RhoE|Rnd3|RND3_HUMAN|Small GTP binding protein Rho8
- ARHE|memB|Protein MemB|Ras homolog gene family member E|Rho family GTPase 3|Rho-related GTP-binding protein Rho8|Rho-related GTP-binding protein RhoE|Rho8|RhoE|Rnd3|RND3_HUMAN|Small GTP binding protein Rho8
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P61587
- P61587
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together