Anti-ITPRID2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ITPRID2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77801-104
- 77801-103
- Antigen symbol:
- ITPRID2
- ITPRID2
- Antigen name:
- Protein ITPRID2 protein (1-267AA)
- Protein ITPRID2 protein (1-267AA)
- Antigen synonyms:
- Cleavage signal-1 protein|CS-1|CS1|Ki-ras-induced actin-interacting protein|KIAA1927|KRAP|KRAS induced actin interacting protein|SPAG13|Sperm associated antigen 13|Sperm-specific antigen 2|SSFA2|SSFA2_HUMAN
- Cleavage signal-1 protein|CS-1|CS1|Ki-ras-induced actin-interacting protein|KIAA1927|KRAP|KRAS induced actin interacting protein|SPAG13|Sperm associated antigen 13|Sperm-specific antigen 2|SSFA2|SSFA2_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P28290
- P28290
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together