Anti-TRPC1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TRPC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-595
- 77803-595
- Antigen symbol:
- TRPC1
- TRPC1
- Antigen name:
- Short transient receptor potential channel 1 protein (39-207AA)
- Short transient receptor potential channel 1 protein (39-207AA)
- Antigen synonyms:
- HTRP 1|HTRP1|MGC133334|MGC133335|Mtrp1|Short transient receptor potential channel 1|Transient receptor potential canonical 1|Transient receptor potential cation channel subfamily C member 1|Transient receptor potential channel 1|Transient receptor protein 1|TRP 1|TRP 1 protein|Trp related protein 1|TRP-1|TRP1|TRP1 protein|TrpC1|Trpc1 transient receptor potential cation channel subfamily C member 1|TRPC1_HUMAN|Trrp1
- HTRP 1|HTRP1|MGC133334|MGC133335|Mtrp1|Short transient receptor potential channel 1|Transient receptor potential canonical 1|Transient receptor potential cation channel subfamily C member 1|Transient receptor potential channel 1|Transient receptor protein 1|TRP 1|TRP 1 protein|Trp related protein 1|TRP-1|TRP1|TRP1 protein|TrpC1|Trpc1 transient receptor potential cation channel subfamily C member 1|TRPC1_HUMAN|Trrp1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P48995
- P48995
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together