Anti-UCHL5 Rabbit Polyclonal Antibody
Catalog # 77811-926
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:UCHL5_HUMAN
- Antigen symbol:UCHL5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:100 µl
- Form:Liquid
- Gene ID:Q9Y5K5
- Antigen synonyms:UCH37|AD-019|CGI 70|INO80 complex subunit R|INO80R|Ubiquitin C terminal hydrolase L5|Ubiquitin C terminal hydrolase UCH37|Ubiquitin C-terminal hydrolase UCH37|Ubiquitin carboxyl terminal esterase L5|Ubiquitin carboxyl terminal hydrolase isozyme L5|Ubiquitin carboxyl terminal hydrolase L5|Ubiquitin carboxyl-terminal hydrolase isozyme L5|Ubiquitin thioesterase L5|Ubiquitin thiolesterase L5|UCH L5|UCH-L5|UCHL5_HUMAN
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: UCHL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat