Anti-COPS7A Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: COPS7A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-188
- 77840-187
- Antigen symbol:
- COPS7A
- COPS7A
- Antigen name:
- COP9 signalosome complex subunit 7a protein (2-275AA)
- COP9 signalosome complex subunit 7a protein (2-275AA)
- Antigen synonyms:
- COP9 complex subunit 7a|COP9 constitutive photomorphogenic homolog subunit 7A|COP9 signalosome complex subunit 7a|COP9 signalosome subunit 7A|COPS7A|CSN7A|CSN7A_HUMAN|Dermal papilla derived protein 10|Dermal papilla-derived protein 10|DERP10|JAB1 containing signalosome subunit 7a|JAB1-containing signalosome subunit 7a|SGN7a|Signalosome subunit 7a
- COP9 complex subunit 7a|COP9 constitutive photomorphogenic homolog subunit 7A|COP9 signalosome complex subunit 7a|COP9 signalosome subunit 7A|COPS7A|CSN7A|CSN7A_HUMAN|Dermal papilla derived protein 10|Dermal papilla-derived protein 10|DERP10|JAB1 containing signalosome subunit 7a|JAB1-containing signalosome subunit 7a|SGN7a|Signalosome subunit 7a
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UBW8
- Q9UBW8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together