Anti-RGS22 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: RGS22
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77797-222
- 77797-221
- Antigen symbol:
- RGS22
- RGS22
- Antigen name:
- Regulator of G-protein signaling 22 protein (201-504AA)
- Regulator of G-protein signaling 22 protein (201-504AA)
- Antigen synonyms:
- FLJ40080|FLJ75004|MGC102908|PRTD-NY2|Regulator of G-protein signaling 22|RGS22|RGS22_HUMAN
- FLJ40080|FLJ75004|MGC102908|PRTD-NY2|Regulator of G-protein signaling 22|RGS22|RGS22_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NE09
- Q8NE09
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together