Anti-EIF3G Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: EIF3G
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77806-481
- 77806-480
- Antigen symbol:
- EIF3G
- EIF3G
- Antigen name:
- Eukaryotic translation initiation factor 3 subunit G protein (2-320AA)
- Eukaryotic translation initiation factor 3 subunit G protein (2-320AA)
- Antigen synonyms:
- 44kDa|Anti EIF3S4|eIF 3 delta|eIF 3 RNA binding subunit|eIF-3 RNA-binding subunit|eIF-3-delta|eIF3 delta|eIF3 p42|eIF3 p44|EIF3G_HUMAN|EIF3S4|formerly|Eukaryotic translation initiation factor 3|Eukaryotic translation initiation factor 3 RNA binding subunit|Eukaryotic translation initiation factor 3 RNA-binding subunit|Eukaryotic translation initiation factor 3 subunit 4|Eukaryotic translation initiation factor 3 subunit 4 delta 44 kD|Eukaryotic translation initiation factor 3 subunit G|Eukaryotic translation initiation factor 3 subunit p42|subunit 4|Subunit 4 (delta 44kD)|Subunit 4 delta
- 44kDa|Anti EIF3S4|eIF 3 delta|eIF 3 RNA binding subunit|eIF-3 RNA-binding subunit|eIF-3-delta|eIF3 delta|eIF3 p42|eIF3 p44|EIF3G_HUMAN|EIF3S4|formerly|Eukaryotic translation initiation factor 3|Eukaryotic translation initiation factor 3 RNA binding subunit|Eukaryotic translation initiation factor 3 RNA-binding subunit|Eukaryotic translation initiation factor 3 subunit 4|Eukaryotic translation initiation factor 3 subunit 4 delta 44 kD|Eukaryotic translation initiation factor 3 subunit G|Eukaryotic translation initiation factor 3 subunit p42|subunit 4|Subunit 4 (delta 44kD)|Subunit 4 delta
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75821
- O75821
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together