Anti-VPS25 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:2000 to 1:5000, WB: 1:500 to 1:2000
Type:
Antigen: VPS25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77818-380
- 77818-379
- Antigen symbol:
- VPS25
- VPS25
- Antigen name:
- Full length fusion protein
- Full length fusion protein
- Antigen synonyms:
- Dermal papilla derived protein 9|Dermal papilla-derived protein 9|DERP9|EAP20|ELL associated protein of 20 kDa|ELL-associated protein of 20 kDa|ESCRT-II complex subunit VPS25|FAP20|HGNC:28122|hVps25|MGC10540|Vacuolar protein sorting 25 homolog (S. cerevisiae)|Vacuolar protein sorting associated protein 25|Vacuolar protein-sorting-associated protein 25|VPS25|VPS25_HUMAN
- Dermal papilla derived protein 9|Dermal papilla-derived protein 9|DERP9|EAP20|ELL associated protein of 20 kDa|ELL-associated protein of 20 kDa|ESCRT-II complex subunit VPS25|FAP20|HGNC:28122|hVps25|MGC10540|Vacuolar protein sorting 25 homolog (S. cerevisiae)|Vacuolar protein sorting associated protein 25|Vacuolar protein-sorting-associated protein 25|VPS25|VPS25_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BRG1
- Q9BRG1
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together