Anti-RPLP2 Rabbit Polyclonal Antibody
Catalog # 77807-405
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Synthesized peptide derived from the Internal region of Ribosomal Protein LP2.
- Antigen symbol:RPLP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:50 µg
- Form:Liquid
- Gene ID:P05387
- Antigen synonyms:2700049I22Rik|60S acidic ribosomal protein P2|Acidic ribosomal phosphoprotein P2|D11S2243E|LP2|MGC71408|OTTMUSP00000029428|OTTMUSP00000029429|OTTMUSP00000029430|P2|Renal carcinoma antigen NY-REN-44|Ribosomal protein P2|Ribosomal protein, large, P2|RLA2_HUMAN|rplP2|RPP2
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:40000
Type:
Antigen: RPLP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat