Anti-PSMB8 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PSMB8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77795-803
- 77795-804
- Antigen symbol:
- PSMB8
- PSMB8
- Antigen name:
- Proteasome subunit beta type-8 protein (176-276AA)
- Proteasome subunit beta type-8 protein (176-276AA)
- Antigen synonyms:
- ALDD| D6S216| D6S216E| Large multifunctional peptidase 7| Large multifunctional protease 7| LMP 7| LMP7| Low molecular mass protein 7| Low molecular weight protein 7| Macropain subunit C13| MGC1491| Multicatalytic endopeptidase complex subunit C13| NKJO| OTTHUMP00000062981| Protease component C13| Proteasome (prosome macropain) subunit beta type 8| Proteasome (prosome, macropain) subunit, beta type, 8 (large multifunctional peptidase 7)| Proteasome beta 8 subunit| Proteasome catalytic subunit 3i| Proteasome component C13| Proteasome related gene 7| Proteasome subunit beta 5i| Proteasome subunit beta 8| Proteasome subunit beta type 8| Proteasome subunit beta type| Proteasome subunit beta type-8| Proteasome subunit beta-5i| Proteasome subunit Y2| PSB8_HUMAN| PSMB 8| PSMB5i| PSMB8| Really interesting new gene 10 protein| RING 10| RING10| Y2
- ALDD| D6S216| D6S216E| Large multifunctional peptidase 7| Large multifunctional protease 7| LMP 7| LMP7| Low molecular mass protein 7| Low molecular weight protein 7| Macropain subunit C13| MGC1491| Multicatalytic endopeptidase complex subunit C13| NKJO| OTTHUMP00000062981| Protease component C13| Proteasome (prosome macropain) subunit beta type 8| Proteasome (prosome, macropain) subunit, beta type, 8 (large multifunctional peptidase 7)| Proteasome beta 8 subunit| Proteasome catalytic subunit 3i| Proteasome component C13| Proteasome related gene 7| Proteasome subunit beta 5i| Proteasome subunit beta 8| Proteasome subunit beta type 8| Proteasome subunit beta type| Proteasome subunit beta type-8| Proteasome subunit beta-5i| Proteasome subunit Y2| PSB8_HUMAN| PSMB 8| PSMB5i| PSMB8| Really interesting new gene 10 protein| RING 10| RING10| Y2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P28062
- P28062
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together