Anti-luxS Rabbit Recombinant Polyclonal Antibody
Catalog # 77819-681
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Recombinant Escherichia coli S-ribosylhomocysteine lyase protein (2-171AA)
- Antigen symbol:luxS
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:E. coli
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P45578
- Antigen synonyms:luxS|ygaG|b2687|JW2662|S-ribosylhomocysteine lyase|EC 4.4.1.21|AI-2 synthesis protein|Autoinducer-2 production protein LuxS
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000
Type:
Antigen: luxS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Escherichia coli