Anti-ZFPM2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: ZFPM2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-664
- 77833-663
- Antigen symbol:
- ZFPM2
- ZFPM2
- Antigen name:
- Zinc finger protein ZFPM2 protein (982-1151AA)
- Zinc finger protein ZFPM2 protein (982-1151AA)
- Antigen synonyms:
- FOG-2|FOG2_HUMAN|Friend of GATA 2|Friend of GATA protein 2|Friend of GATA2|hFOG-2|Zinc finger protein 89B|Zinc finger protein M2|Zinc finger protein multitype 2|Zinc finger protein ZFPM2
- FOG-2|FOG2_HUMAN|Friend of GATA 2|Friend of GATA protein 2|Friend of GATA2|hFOG-2|Zinc finger protein 89B|Zinc finger protein M2|Zinc finger protein multitype 2|Zinc finger protein ZFPM2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WW38
- Q8WW38
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together