Anti-UQCRC2 Rabbit Polyclonal Antibody
Catalog # 77804-064
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Complex III subunit 2
- Antigen symbol:UQCRC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P22695
- Antigen synonyms:Complex III subunit 2|Core protein II|Cytochrome b c1 complex subunit 2 mitochondrial|Cytochrome b-c1 complex subunit 2|EC 1.10.2.2|MC3DN5|mitochondrial|QCR2|QCR2_HUMAN|Ubiquinol cytochrome c reductase complex core protein 2 mitochondrial|Ubiquinol cytochrome c reductase core protein II|Ubiquinol-cytochrome-c reductase complex core protein 2|UQCR2|Uqcrc2
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: UQCRC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat