Anti-POLD1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: POLD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77794-682
- 77794-683
- Antigen symbol:
- POLD1
- POLD1
- Antigen name:
- DNA polymerase delta catalytic subunit protein (1-123AA)
- DNA polymerase delta catalytic subunit protein (1-123AA)
- Antigen synonyms:
- Polymerase (DNA directed) delta 1 catalytic subunit|CDC2|CDC2 homolog|CRCS10|DNA directed DNA polymerase delta 1|DNA directed polymerase delta 1|DNA pol delta 1|DNA polymerase delta catalytic subunit|DNA polymerase subunit delta p125|DPOD1_HUMAN|MDPL|POLD|POLD 1|POLD1|Polymerase (DNA directed) delta 1 catalytic subunit 125kDa|Polymerase (DNA) delta 1 catalytic subunit|Polymerase DNA directed delta 1 catalytic subunit 125kD|polymerase, DNA, delta
- Polymerase (DNA directed) delta 1 catalytic subunit|CDC2|CDC2 homolog|CRCS10|DNA directed DNA polymerase delta 1|DNA directed polymerase delta 1|DNA pol delta 1|DNA polymerase delta catalytic subunit|DNA polymerase subunit delta p125|DPOD1_HUMAN|MDPL|POLD|POLD 1|POLD1|Polymerase (DNA directed) delta 1 catalytic subunit 125kDa|Polymerase (DNA) delta 1 catalytic subunit|Polymerase DNA directed delta 1 catalytic subunit 125kD|polymerase, DNA, delta
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P28340
- P28340
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together