Anti-SLC37A1 Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: SLC37A1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77800-057
- 77800-058
- Antigen symbol:
- SLC37A1
- SLC37A1
- Antigen name:
- Glucose-6-phosphate exchanger SLC37A1 protein (245-302AA)
- Glucose-6-phosphate exchanger SLC37A1 protein (245-302AA)
- Antigen synonyms:
- G-3-P permease|G-3-P transporter|G3P permease|G3P transporter|G3PP|GLPT_HUMAN|Glycerol-3-phosphate permease|Glycerol-3-phosphate transporter|SLC37A1|solute carrier family 37 (glycerol-3-phosphate transporter)|member 1|Solute carrier family 37 member 1
- G-3-P permease|G-3-P transporter|G3P permease|G3P transporter|G3PP|GLPT_HUMAN|Glycerol-3-phosphate permease|Glycerol-3-phosphate transporter|SLC37A1|solute carrier family 37 (glycerol-3-phosphate transporter)|member 1|Solute carrier family 37 member 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P57057
- P57057
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together