Anti-FHL1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FHL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-931
- 77823-930
- Antigen symbol:
- FHL1
- FHL1
- Antigen name:
- Four and a half LIM domains protein 1 protein (13-240AA)
- Four and a half LIM domains protein 1 protein (13-240AA)
- Antigen synonyms:
- bA535K18.1|FHL 1|FHL 1B|FHL-1|FHL1|FHL1 protein|FHL1_HUMAN|FHL1A|FHL1B|FLH1A|Four and a half LIM domains 1|Four and a half LIM domains protein 1|Four and a half Lin11 Isl 1 and Mec 3 domains 1|four-and-a-half Lin11|Isl-1 and Mec-3|KYO T|KYOT|mouse|homolog of|LIM protein SLIMMER|MGC111107|RAM14-1|RBMX1A|RBMX1B|RBP associated molecule 14-1|RP11-535K18.1|Skeletal muscle LIM protein 1|Skeletal muscle LIM-protein 1|SLIM 1|SLIM|SLIM-1|SLIM1|SLIMMER|XMPMA
- bA535K18.1|FHL 1|FHL 1B|FHL-1|FHL1|FHL1 protein|FHL1_HUMAN|FHL1A|FHL1B|FLH1A|Four and a half LIM domains 1|Four and a half LIM domains protein 1|Four and a half Lin11 Isl 1 and Mec 3 domains 1|four-and-a-half Lin11|Isl-1 and Mec-3|KYO T|KYOT|mouse|homolog of|LIM protein SLIMMER|MGC111107|RAM14-1|RBMX1A|RBMX1B|RBP associated molecule 14-1|RP11-535K18.1|Skeletal muscle LIM protein 1|Skeletal muscle LIM-protein 1|SLIM 1|SLIM|SLIM-1|SLIM1|SLIMMER|XMPMA
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13642
- Q13642
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together