Anti-HLA-E Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HLA-E
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77815-198
- 77815-197
- Antigen symbol:
- HLA-E
- HLA-E
- Antigen name:
- HLA class I histocompatibility antigen, alpha chain E protein (33-261AA)
- HLA class I histocompatibility antigen, alpha chain E protein (33-261AA)
- Antigen synonyms:
- HLA-6.2|HLAEHLA class I histocompatibility antigen|alpha chain E|MHC class I antigen E) [Cleaved into: Soluble HLA class I histocompatibility antigen|sHLA-E)]
- HLA-6.2|HLAEHLA class I histocompatibility antigen|alpha chain E|MHC class I antigen E) [Cleaved into: Soluble HLA class I histocompatibility antigen|sHLA-E)]
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P13747
- P13747
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together