Anti-YIF1B Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: YIF1B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77827-005
- 77827-006
- Antigen symbol:
- YIF1B
- YIF1B
- Antigen name:
- Protein YIF1B protein (1-153AA)
- Protein YIF1B protein (1-153AA)
- Antigen synonyms:
- FinGER8|FLJ50074|Protein YIF1B|YIF1B|YIF1B_HUMAN|Yip1 interacting factor homolog B (S. cerevisiae)|Yip1 interacting factor homolog B|YIP1-interacting factor homolog B|Yip1b
- FinGER8|FLJ50074|Protein YIF1B|YIF1B|YIF1B_HUMAN|Yip1 interacting factor homolog B (S. cerevisiae)|Yip1 interacting factor homolog B|YIP1-interacting factor homolog B|Yip1b
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5BJH7
- Q5BJH7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together