Anti-UBE2L3 Rabbit Polyclonal Antibody
Catalog # 77803-861
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Ubiquitin-protein ligase L3
- Antigen symbol:UBE2L3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P68036
- Antigen synonyms:E2 F1|L UBC|L-UBC|UB2L3_HUMAN|UBCE7|UbcH7|UbcM4|Ubiquitin carrier protein L3|Ubiquitin conjugating enzyme E2 L3|Ubiquitin protein ligase L3|Ubiquitin-conjugating enzyme E2 L3|Ubiquitin-conjugating enzyme E2-F1|Ubiquitin-protein ligase L3
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: UBE2L3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat