Anti-AGXT2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: AGXT2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77836-926
- 77836-927
- Antigen symbol:
- AGXT2
- AGXT2
- Antigen name:
- Alanine--glyoxylate aminotransferase 2, mitochondrial protein (196-328AA)
- Alanine--glyoxylate aminotransferase 2, mitochondrial protein (196-328AA)
- Antigen synonyms:
- AGXT2|AGT2Alanine--glyoxylate aminotransferase 2|mitochondrial|AGT 2|EC 2.6.1.44|R)-3-amino-2-methylpropionate--pyruvate transaminase|EC 2.6.1.40|Beta-ALAAT II|Beta-alanine-pyruvate aminotransferase|D-AIBAT
- AGXT2|AGT2Alanine--glyoxylate aminotransferase 2|mitochondrial|AGT 2|EC 2.6.1.44|R)-3-amino-2-methylpropionate--pyruvate transaminase|EC 2.6.1.40|Beta-ALAAT II|Beta-alanine-pyruvate aminotransferase|D-AIBAT
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BYV1
- Q9BYV1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together