Anti-TXNL4B Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: TXNL4B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77837-565
- 77837-564
- Antigen symbol:
- TXNL4B
- TXNL4B
- Antigen name:
- Thioredoxin-like protein 4B protein (1-149AA)
- Thioredoxin-like protein 4B protein (1-149AA)
- Antigen synonyms:
- AI595343|D530025J19Rik|Dim1 like protein|Dim1-like protein|DIM2|DLP|FLJ20511|MGC109322|RGD1305127|Thioredoxin like 4B|Thioredoxin like protein 4B|Thioredoxin-like protein 4B|TXN4B_HUMAN|Txnl4b
- AI595343|D530025J19Rik|Dim1 like protein|Dim1-like protein|DIM2|DLP|FLJ20511|MGC109322|RGD1305127|Thioredoxin like 4B|Thioredoxin like protein 4B|Thioredoxin-like protein 4B|TXN4B_HUMAN|Txnl4b
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NX01
- Q9NX01
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together