Anti-MYO10 Rabbit Recombinant Polyclonal Antibody (FITC)
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
Type:
Antigen: MYO10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spécifications
- Catalog No:
- 77842-237
- 77842-238
- Antigen symbol:
- MYO10
- MYO10
- Antigen name:
- Unconventional myosin-X protein (1-97AA)
- Unconventional myosin-X protein (1-97AA)
- Antigen synonyms:
- AW048724|D15Ertd600e|FLJ10639|FLJ21066|FLJ22268|FLJ43256|KIAA0799|MGC131988|mKIAA0799|MYO10|MYO10_HUMAN|Myosin X|Myosin-X|OTTMUSP00000022066|Unconventional myosin 10|Unconventional myosin X|Unconventional myosin-10
- AW048724|D15Ertd600e|FLJ10639|FLJ21066|FLJ22268|FLJ43256|KIAA0799|MGC131988|mKIAA0799|MYO10|MYO10_HUMAN|Myosin X|Myosin-X|OTTMUSP00000022066|Unconventional myosin 10|Unconventional myosin X|Unconventional myosin-10
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HD67
- Q9HD67
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Spécifications
Frequently Bought Together