Anti-SMAD7 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:5000, IF: 1:20 to 1:200
Type:
Antigen: SMAD7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77800-265
- 77800-264
- Antigen symbol:
- SMAD7
- SMAD7
- Antigen name:
- Mothers against decapentaplegic homolog 7 protein (36-247AA)
- Mothers against decapentaplegic homolog 7 protein (36-247AA)
- Antigen synonyms:
- CRCS3|FLJ16482|hSMAD 7|hSMAD7|MAD (mothers against decapentaplegic Drosophila) homolog 7|MAD|Mad homolog 7|MAD homolog 8|MAD mothers against decapentaplegic homolog 7|MADH 7|MADH 8|MADH6|MADH7|MADH8|Mothers Against Decapentaplegic Drosophila Homolog of 6|Mothers Against Decapentaplegic Drosophila Homolog of 7|Mothers against decapentaplegic homolog 7|Mothers against decapentaplegic homolog 8|Mothers against DPP homolog 7|Mothers against DPP homolog 8|SMA- AND MAD-RELATED PROTEIN 7|SMAD 7|SMAD|SMAD family member 7|SMAD, mothers against DPP homolog 7 (Drosophila)|SMAD, mothers against DPP homolog 7|SMAD6|Smad7|SMAD7_HUMAN
- CRCS3|FLJ16482|hSMAD 7|hSMAD7|MAD (mothers against decapentaplegic Drosophila) homolog 7|MAD|Mad homolog 7|MAD homolog 8|MAD mothers against decapentaplegic homolog 7|MADH 7|MADH 8|MADH6|MADH7|MADH8|Mothers Against Decapentaplegic Drosophila Homolog of 6|Mothers Against Decapentaplegic Drosophila Homolog of 7|Mothers against decapentaplegic homolog 7|Mothers against decapentaplegic homolog 8|Mothers against DPP homolog 7|Mothers against DPP homolog 8|SMA- AND MAD-RELATED PROTEIN 7|SMAD 7|SMAD|SMAD family member 7|SMAD, mothers against DPP homolog 7 (Drosophila)|SMAD, mothers against DPP homolog 7|SMAD6|Smad7|SMAD7_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15105
- O15105
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together