Anti-NTM Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: NTM
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-305
- 77840-306
- Antigen symbol:
- NTM
- NTM
- Antigen name:
- Neurotrimin protein (30-316AA)
- Neurotrimin protein (30-316AA)
- Antigen synonyms:
- hNT|IgLON family member 2|IGLON2|Neurotrimin|NT|NTM|NTRI_HUMAN
- hNT|IgLON family member 2|IGLON2|Neurotrimin|NT|NTM|NTRI_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9P121
- Q9P121
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together