Anti-MAIP1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MAIP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-670
- 77833-671
- Antigen symbol:
- MAIP1
- MAIP1
- Antigen name:
- m-AAA protease-interacting protein 1, mitochondrial protein (97-291AA)
- m-AAA protease-interacting protein 1, mitochondrial protein (97-291AA)
- Antigen synonyms:
- C2orf47|mitochondrial|CB047_HUMAN|Chromosome 2 open reading frame 47|Hhypothetical protein LOC79568|m AAA protease interacting protein 1|uncharacterized protein|Matrix AAA peptidase interacting protein 1|Uncharacterized protein C2orf47
- C2orf47|mitochondrial|CB047_HUMAN|Chromosome 2 open reading frame 47|Hhypothetical protein LOC79568|m AAA protease interacting protein 1|uncharacterized protein|Matrix AAA peptidase interacting protein 1|Uncharacterized protein C2orf47
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WWC4
- Q8WWC4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together