Anti-SENP1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:50 to 1:200
Type:
Antigen: SENP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77806-023
- 77806-024
- Antigen symbol:
- SENP1
- SENP1
- Antigen name:
- Synthetic peptide of SENP1
- Synthetic peptide of SENP1
- Antigen synonyms:
- SENP 1|SENP1|SENP1_HUMAN|Sentrin specific protease 1|Sentrin-specific protease 1|Sentrin/SUMO specific protease 1|Sentrin/SUMO specific protease|Sentrin/SUMO specific protease SENP 1|Sentrin/SUMO specific protease SENP1|Sentrin/SUMO-specific protease SENP1|SUMO1/sentrin specific peptidase 1|SUMO1/sentrin specific protease 1|SuPr 2|SuPr2
- SENP 1|SENP1|SENP1_HUMAN|Sentrin specific protease 1|Sentrin-specific protease 1|Sentrin/SUMO specific protease 1|Sentrin/SUMO specific protease|Sentrin/SUMO specific protease SENP 1|Sentrin/SUMO specific protease SENP1|Sentrin/SUMO-specific protease SENP1|SUMO1/sentrin specific peptidase 1|SUMO1/sentrin specific protease 1|SuPr 2|SuPr2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9P0U3
- Q9P0U3
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together