Anti-atpE Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: atpE
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: strain OhILAs
Specifications
- Catalog No:
- 77818-257
- 77818-256
- Antigen symbol:
- atpE
- atpE
- Antigen name:
- Recombinant strain OhILAs ATP synthase subunit c protein (1-88AA)
- Recombinant strain OhILAs ATP synthase subunit c protein (1-88AA)
- Antigen synonyms:
- atpE|Clos_2565|ATP synthase subunit c|ATP synthase F(0) sector subunit c|F-type ATPase subunit c|F-ATPase subunit c|Lipid-binding protein
- atpE|Clos_2565|ATP synthase subunit c|ATP synthase F(0) sector subunit c|F-type ATPase subunit c|F-ATPase subunit c|Lipid-binding protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- OhILAs strain
- OhILAs strain
- Host:
- Rabbit
- Rabbit
- Gene ID:
- A8MJW4
- A8MJW4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together