Anti-FARSB Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FARSB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-968
- 77840-969
- Antigen symbol:
- FARSB
- FARSB
- Antigen name:
- Phenylalanine--tRNA ligase beta subunit protein (1-299AA)
- Phenylalanine--tRNA ligase beta subunit protein (1-299AA)
- Antigen synonyms:
- C76708|Farsa|FARSB|Farsl|Farslb|FRSB|HSPC173|PheHB|Phenylalanine tRNA ligase beta subunit|phenylalanine tRNA ligase 1, beta, cytoplasmic|Phenylalanine tRNA ligase beta chain|Phenylalanine--tRNA ligase beta chain|phenylalanine-tRNA synthetase-like, beta subunit|Phenylalanyl tRNA synthetase beta chain|Phenylalanyl-tRNA synthetase beta chain|phenylalanyl-tRNA synthetase beta-subunit|phenylalanyl-tRNA synthetase-like, beta subunit|PheRS|SYFB_HUMAN
- C76708|Farsa|FARSB|Farsl|Farslb|FRSB|HSPC173|PheHB|Phenylalanine tRNA ligase beta subunit|phenylalanine tRNA ligase 1, beta, cytoplasmic|Phenylalanine tRNA ligase beta chain|Phenylalanine--tRNA ligase beta chain|phenylalanine-tRNA synthetase-like, beta subunit|Phenylalanyl tRNA synthetase beta chain|Phenylalanyl-tRNA synthetase beta chain|phenylalanyl-tRNA synthetase beta-subunit|phenylalanyl-tRNA synthetase-like, beta subunit|PheRS|SYFB_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NSD9
- Q9NSD9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together