Anti-CHST3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CHST3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77831-076
- 77831-077
- Antigen symbol:
- CHST3
- CHST3
- Antigen name:
- Carbohydrate sulfotransferase 3 protein (316-427AA)
- Carbohydrate sulfotransferase 3 protein (316-427AA)
- Antigen synonyms:
- C6ST 1|C6ST|C6ST-1|C6ST1|Carbohydrate (chondroitin 6) sulfotransferase 3|Carbohydrate sulfotransferase 3|Chondroitin 6 O sulfotransferase 1|Chondroitin 6 sulfotransferase|Chondroitin 6-O-sulfotransferase 1|Chondroitin 6-sulfotransferase|CHST 3|Chst3|CHST3_HUMAN|Galactose/N acetylglucosamine/N acetylglucosamine 6 O sulfotransferase 0|Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 0|GST 0|GST-0|GST0
- C6ST 1|C6ST|C6ST-1|C6ST1|Carbohydrate (chondroitin 6) sulfotransferase 3|Carbohydrate sulfotransferase 3|Chondroitin 6 O sulfotransferase 1|Chondroitin 6 sulfotransferase|Chondroitin 6-O-sulfotransferase 1|Chondroitin 6-sulfotransferase|CHST 3|Chst3|CHST3_HUMAN|Galactose/N acetylglucosamine/N acetylglucosamine 6 O sulfotransferase 0|Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 0|GST 0|GST-0|GST0
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7LGC8
- Q7LGC8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together