Anti-TUBB1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77837-175
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Tubulin beta-1 chain protein (270-451AA)
- Antigen symbol:TUBB1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9H4B7
- Antigen synonyms:2810484G07Rik|beta 1 tubulin|Beta tubulin 1, class VI|Beta-tubulin|Class VI beta tubulin|dJ543J19.4|M(beta)1|TBB1_HUMAN|TUBB1|Tubulin beta 1 class VI|Tubulin beta-1 chain|Tubulin, beta 1|tubulin, beta1
- Storage buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TUBB1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human