Anti-LATS2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: LATS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse
Specifications
- Catalog No:
- 77827-313
- 77827-314
- Antigen symbol:
- LATS2
- LATS2
- Antigen name:
- Synthetic peptide of LATS2
- Synthetic peptide of LATS2
- Antigen synonyms:
- FLJ13161|Kinase phosphorylated during mitosis protein|KPM|Large tumor suppressor homolog 2|Large tumor suppressor homolog 2 Drosophila|Large tumor suppressor kinase 2|LATS large tumor suppressor Drosophila homolog 2|LATS large tumor suppressor homolog 2|Lats2|LATS2_HUMAN|Serine/threonine kinase kpm|Serine/threonine protein kinase kpm|Serine/threonine-protein kinase kpm|Serine/threonine-protein kinase LATS2|Warts like kinase|Warts-like kinase
- FLJ13161|Kinase phosphorylated during mitosis protein|KPM|Large tumor suppressor homolog 2|Large tumor suppressor homolog 2 Drosophila|Large tumor suppressor kinase 2|LATS large tumor suppressor Drosophila homolog 2|LATS large tumor suppressor homolog 2|Lats2|LATS2_HUMAN|Serine/threonine kinase kpm|Serine/threonine protein kinase kpm|Serine/threonine-protein kinase kpm|Serine/threonine-protein kinase LATS2|Warts like kinase|Warts-like kinase
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NRM7
- Q9NRM7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together