Anti-MRM3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MRM3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77837-301
- 77837-302
- Antigen symbol:
- MRM3
- MRM3
- Antigen name:
- rRNA methyltransferase 3, mitochondrial protein (41-205AA)
- rRNA methyltransferase 3, mitochondrial protein (41-205AA)
- Antigen synonyms:
- 16S rRNA (guanosine(1370)-2'-O)-methyltransferase|16S rRNA [Gm1370] 2'-O-methyltransferase|4833420N02Rik|AI255153|FLJ10581|HC90|MRM3|MRM3_HUMAN|Putative RNA methyltransferase|RMTL1|RNA methyltransferase like 1|RNA methyltransferase like protein 1|RNA methyltransferase-like protein 1|RP23-147P4.7|rRNA methyltransferase 3, mitochondrial
- 16S rRNA (guanosine(1370)-2'-O)-methyltransferase|16S rRNA [Gm1370] 2'-O-methyltransferase|4833420N02Rik|AI255153|FLJ10581|HC90|MRM3|MRM3_HUMAN|Putative RNA methyltransferase|RMTL1|RNA methyltransferase like 1|RNA methyltransferase like protein 1|RNA methyltransferase-like protein 1|RP23-147P4.7|rRNA methyltransferase 3, mitochondrial
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HC36
- Q9HC36
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together