Anti-RPL35A Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: RPL35A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77797-977
- 77797-976
- Antigen symbol:
- RPL35A
- RPL35A
- Antigen name:
- 60S ribosomal protein L35a (1-110AA)
- 60S ribosomal protein L35a (1-110AA)
- Antigen synonyms:
- 60S ribosomal protein L35a|Cell growth-inhibiting gene 33 protein|DBA5|L35A|Ribosomal Protein L35a|RL35A_HUMAN|RPL35A
- 60S ribosomal protein L35a|Cell growth-inhibiting gene 33 protein|DBA5|L35A|Ribosomal Protein L35a|RL35A_HUMAN|RPL35A
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P18077
- P18077
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together