Anti-BAG6 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: BAG6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77816-393
- 77816-392
- Antigen symbol:
- BAG6
- BAG6
- Antigen name:
- Large proline-rich protein BAG6 protein (1-200AA)
- Large proline-rich protein BAG6 protein (1-200AA)
- Antigen synonyms:
- 2410045D21Rik|AA408914|BAG 6|BAG family molecular chaperone regulator 6|BAG-6|BAG6|BAG6_HUMAN|BAT 3|BAT3|BCL2-associated athanogene 6|D17H6S52E|D6S52E|G3|HLA B associated transcript 3|HLA-B associated transcript 3|HLA-B associated transcript-3|HLA-B-associated transcript 3|large proline rich protein BAG6|Large proline rich protein BAT3|Large proline-rich protein BAG6|large proline-rich protein BAT3|Protein G3|Protein Scythe|Scythe|homolog of Xenopus
- 2410045D21Rik|AA408914|BAG 6|BAG family molecular chaperone regulator 6|BAG-6|BAG6|BAG6_HUMAN|BAT 3|BAT3|BCL2-associated athanogene 6|D17H6S52E|D6S52E|G3|HLA B associated transcript 3|HLA-B associated transcript 3|HLA-B associated transcript-3|HLA-B-associated transcript 3|large proline rich protein BAG6|Large proline rich protein BAT3|Large proline-rich protein BAG6|large proline-rich protein BAT3|Protein G3|Protein Scythe|Scythe|homolog of Xenopus
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P46379
- P46379
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together